2,5-Dichloro-4-iodopyridine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR345414-10G OR345414-25G OR345414-5G
APOSOR345414-10GEA 128 GBP
APOSOR345414-10G APOSOR345414-25G APOSOR345414-5G
2,5-Dichloro-4-iodopyridine 98%
2,5-Dichloro-4-iodopyridine
Formula: C₅H₂Cl₂IN
Storage Temperature: Ambient
MDL Number: MFCD13185538
CAS Number: 796851-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR