3-Bromo-(5-methoxycarbonyl)phenylboronic acid 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Bromo-5-(methoxycarbonyl)phenylboronic acid, (3-Bromo-5-(methoxycarbonyl)phenyl)boronic acid

OR3434-1G
APOSOR3434-1GEA 52.5 GBP
APOSOR3434-1G
3-Bromo-(5-methoxycarbonyl)phenylboronic acid 96%
3-Bromo-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BBrO₄
MW: 258.8617 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689544
CAS Number: 913835-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR