(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR340028-250MG
APOSOR340028-250MGEA 221 GBP
APOSOR340028-250MG
(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formula: C₃₂H₁₇Cl₄O₄P
Storage Temperature: Refrigerator
MDL Number: MFCD23698720
CAS Number: 1191451-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR