tert-Butyl tetrahydro-1(2H)-pyridazinecarboxylate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: tert-Butyl 1,2-diazinane-1-carboxylate, tert-Butyl tetrahydropyridazine-1(2H)-carboxylate

OR33510-1G
APOSOR33510-1GEA 632 GBP
APOSOR33510-1G
tert-Butyl tetrahydro-1(2H)-pyridazinecarboxylate 95%
tert-Butyl tetrahydro-1(2H)-pyridazinecarboxylate
Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09152721
CAS Number: 154972-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR