2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33256-1G OR33256-5G
APOSOR33256-1GEA 88.1 GBP
APOSOR33256-1G APOSOR33256-5G
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile 95%
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile
Formula: C₁₂H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD01443818
CAS Number: 339105-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR