4-Chloro-2-methyl-1H-1,3-benzodiazole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33139-1G OR33139-5G
APOSOR33139-1GEA 245 GBP
APOSOR33139-1G APOSOR33139-5G
4-Chloro-2-methyl-1H-1,3-benzodiazole 95%
4-Chloro-2-methyl-1H-1,3-benzodiazole
Formula: C₈H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD02572038
CAS Number: 5599-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR