4,6-Dimethyl-3,4-dihydro-2H-1-benzothiopyran-4-ol, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33028-1G
APOSOR33028-1GEA 1140 GBP
APOSOR33028-1G
4,6-Dimethyl-3,4-dihydro-2H-1-benzothiopyran-4-ol, tech
4,6-Dimethyl-3,4-dihydro-2H-1-benzothiopyran-4-ol
Formula: C₁₁H₁₄OS
MW: 194.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02571589
CAS Number: 478065-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR