1-(4-Chlorophenyl)-3-hydroxy-3-methylurea, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32445-1G
APOSOR32445-1GEA 1140 GBP
APOSOR32445-1G
1-(4-Chlorophenyl)-3-hydroxy-3-methylurea, tech
1-(4-Chlorophenyl)-3-hydroxy-3-methylurea
Formula: C₈H₉ClN₂O₂
MW: 200.622 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01814620
CAS Number: 26816-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR