4-Nitrophenol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR322090-25G OR322090-5G OR322090-100G
APOSOR322090-25GEA 17.9 GBP
APOSOR322090-25G APOSOR322090-5G APOSOR322090-100G
4-Nitrophenol 98%
4-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 279 °C
Melting Pt: 114 °C
Density: 1.48 g/cm³ (25 °C)
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
UN: 1663
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR