3-Bromo-4-methoxybenzyl alcohol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (3-Bromo-4-methoxyphenyl)methanol

OR321003-25G
APOSOR321003-25GEA 162 GBP
APOSOR321003-25G
3-Bromo-4-methoxybenzyl alcohol 98%
3-Bromo-4-methoxybenzyl alcohol
Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD01861398
CAS Number: 38493-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR