1-[2-(4-Chlorophenyl)cyclopropyl]ethan-1-one, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32085-1G
APOSOR32085-1GEA 1140 GBP
APOSOR32085-1G
1-[2-(4-Chlorophenyl)cyclopropyl]ethan-1-one, tech
1-[2-(4-Chlorophenyl)cyclopropyl]ethan-1-one
Formula: C₁₁H₁₁ClO
MW: 194.657 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01315475
CAS Number: 54064-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR