7H-Pyrrolo[2,3-d]pyrimidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR317000-1G OR317000-5G OR317000-25G
APOSOR317000-1GEA 20 GBP
APOSOR317000-1G APOSOR317000-5G APOSOR317000-25G
7H-Pyrrolo[2,3-d]pyrimidine
7H-Pyrrolo[2,3-d]pyrimidine
Formula: C₆H₅N₃
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD08234403
CAS Number: 271-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR