2,3-Diaminobut-2-ene-1,4-dinitrile 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2,3-Diamino-1,4-dicyanobut-2-ene

OR3131-25G
APOSOR3131-25GEA 56.7 GBP
APOSOR3131-25G
2,3-Diaminobut-2-ene-1,4-dinitrile 96%
2,3-Diaminobut-2-ene-1,4-dinitrile
Formula: C₄H₄N₄
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00064109
CAS Number: 18514-52-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR