3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Carboxy-3-phenylbicyclo[1.1.1]pentane

OR312133-250MG OR312133-500MG OR312133-100MG
APOSOR312133-250MGEA 256 GBP
APOSOR312133-250MG APOSOR312133-500MG APOSOR312133-100MG
3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid
Formula: C₁₂H₁₂O₂
Melting Pt: 176…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00101889
CAS Number: 83249-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR