(R)-1-Benzyl-5-carboxy-2-pyrrolidinone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR310598-1G
APOSOR310598-1GEA 243 GBP
APOSOR310598-1G
(R)-1-Benzyl-5-carboxy-2-pyrrolidinone
(R)-1-Benzyl-5-carboxy-2-pyrrolidinone
Formula: C₁₂H₁₃NO₃
MW: 219.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797914
CAS Number: 38854-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR