5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR309417-1G
APOSOR309417-1GEA 0 GBP
APOSOR309417-1G
5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole
Formula: C₉H₆Cl₂N₂S
MW: 245.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14707340
CAS Number: 90418-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR