7-Bromo-2,8-dimethyl-4-hydroxyquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR309315-1G
APOSOR309315-1GEA 290 GBP
APOSOR309315-1G
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
Formula: C₁₁H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD12675041
CAS Number: 1189106-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR