2-Amino-5-bromophenol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR30930-1G OR30930-5G OR30930-25G
APOSOR30930-1GEA 15.8 GBP
APOSOR30930-1G APOSOR30930-5G APOSOR30930-25G
2-Amino-5-bromophenol 98%
2-Amino-5-bromophenol
Formula: C₆H₆BrNO
Melting Pt: 135…140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03095028
CAS Number: 38191-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR