6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR309208-1G OR309208-5G
APOSOR309208-1GEA 212 GBP
APOSOR309208-1G APOSOR309208-5G
6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester
6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester
Formula: C₁₃H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD09787817
CAS Number: 114858-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR