6-Bromo-2-chloro-3-methylquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR309201-1G OR309201-5G
APOSOR309201-1GEA 209 GBP
APOSOR309201-1G APOSOR309201-5G
6-Bromo-2-chloro-3-methylquinoline
6-Bromo-2-chloro-3-methylquinoline
Formula: C₁₀H₇BrClN
Storage Temperature: Ambient
MDL Number: MFCD02684154
CAS Number: 113092-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR