Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethyl (R)-1-Boc-3-piperidinecarboxylate, (R)-1-(tert-Butoxycarbonyl)nipecotic Acid Ethyl Ester, (R)-1-Boc-3-piperidinecarboxylic Acid Ethyl Ester, (R)-1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Ethyl Ester, (R)-1-Boc-nipecotic Acid Ethyl Ester, (R)-1-tert-Butyl 3-ethyl piperidine-1,3-dicarboxylate

OR309161-5G
APOSOR309161-5GEA 64.1 GBP
APOSOR309161-5G
Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate
Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate
Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374389
CAS Number: 194726-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR