1-Boc-(2-hydroxymethyl)piperazine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-(Hydroxymethyl)piperazine, N1-BOC protected, 1-Boc-(2-Hydroxymethyl)piperazine

OR308075-250MG
APOSOR308075-250MGEA 15.8 GBP
APOSOR308075-250MG
1-Boc-(2-hydroxymethyl)piperazine 97%
1-Boc-(2-hydroxymethyl)piperazine
Formula: C₁₀H₂₀N₂O₃
MW: 216.2800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772091
CAS Number: 205434-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR