4-Hydrazino-6-ethoxyquinoline hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307973-1G
APOSOR307973-1GEA 271 GBP
APOSOR307973-1G
4-Hydrazino-6-ethoxyquinoline hydrochloride
4-Hydrazino-6-ethoxyquinoline hydrochloride
Formula: C₁₁H₁₃N₃O·ClH
Storage Temperature: Ambient
MDL Number: MFCD09787746
CAS Number: 1172910-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR