2-Chloro-6-ethoxy-3-methylquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307918-1G OR307918-5G
APOSOR307918-1GEA 212 GBP
APOSOR307918-1G APOSOR307918-5G
2-Chloro-6-ethoxy-3-methylquinoline
2-Chloro-6-ethoxy-3-methylquinoline
Formula: C₁₂H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD09787670
CAS Number: 948291-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR