6-Chloro-4-hydrazino-2-methyl-quinoline hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307826-1G
APOSOR307826-1GEA 242 GBP
APOSOR307826-1G
6-Chloro-4-hydrazino-2-methyl-quinoline hydrochloride
6-Chloro-4-hydrazino-2-methyl-quinoline hydrochloride
Formula: C₁₀H₁₀ClN₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD07700274
CAS Number: 1170041-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR