3-(4-aminobenzylamino)-piperidine-1-carboxylic acid tert-butyl ester

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307776-1G
APOSOR307776-1GEA 355 GBP
APOSOR307776-1G
3-(4-aminobenzylamino)-piperidine-1-carboxylic acid tert-butyl ester
3-(4-aminobenzylamino)-piperidine-1-carboxylic acid tert-butyl ester
Formula: C₁₇H₂₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD12674814
CAS Number: 1189105-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR