cis-2-Aminocyclopentanol hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR307607-1G
APOSOR307607-1GEA 625 GBP
APOSOR307607-1G
cis-2-Aminocyclopentanol hydrochloride
cis-2-Aminocyclopentanol hydrochloride
Formula: C₅H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD07366587
CAS Number: 31889-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR