diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide

OR307551-1G OR307551-250MG
APOSOR307551-1GEA 481 GBP
APOSOR307551-1G APOSOR307551-250MG
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
Formula: C₈H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00143157
CAS Number: 105786-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR