Boc-O-methyl-L-serine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

OR307095-1G OR307095-5G
APOSOR307095-1GEA 15.8 GBP
APOSOR307095-1G APOSOR307095-5G
Boc-O-methyl-L-serine
Boc-O-methyl-L-serine
Formula: C₉H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD00153314
CAS Number: 51293-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR