8-Chloroimidazo[1,5-a]pyrazine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR305494-250MG OR305494-100MG OR305494-1G
APOSOR305494-250MGEA 126 GBP
APOSOR305494-250MG APOSOR305494-100MG APOSOR305494-1G
8-Chloroimidazo[1,5-a]pyrazine 95%
8-Chloroimidazo[1,5-a]pyrazine
Formula: C₆H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD20040069
CAS Number: 56468-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR