1,3-Phenylene diisocyanate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR30536-1G OR30536-5G
APOSOR30536-1GEA 49.4 GBP
APOSOR30536-1G APOSOR30536-5G
1,3-Phenylene diisocyanate 98%
1,3-Phenylene diisocyanate

  • May form small amount of sediment.

Formula: C₈H₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00002018
CAS Number: 123-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR