6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR305342-250MG
APOSOR305342-250MGEA 89.8 GBP
APOSOR305342-250MG
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₃ClIN₃
MW: 279.466 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18250995
CAS Number: 1260672-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR