2-Bromo-p-toluic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR3052-100G
APOSOR3052-100GEA 105 GBP
APOSOR3052-100G
2-Bromo-p-toluic acid 97%
2-Bromo-p-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00045797
CAS Number: 7697-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR