tert-Butyl trans-3-aminocyclobutane-1-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR304347-1G
APOSOR304347-1GEA 230 GBP
APOSOR304347-1G
tert-Butyl trans-3-aminocyclobutane-1-carboxylate
tert-Butyl trans-3-aminocyclobutane-1-carboxylate
Formula: C₉H₁₇NO₂
MW: 171.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16661182
CAS Number: 1192547-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR