3-Methoxypyridazine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR304308-5G
APOSOR304308-5GEA 62 GBP
APOSOR304308-5G
3-Methoxypyridazine 97%
3-Methoxypyridazine
Formula: C₅H₆N₂O
MW: 110.1139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234098
CAS Number: 19064-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR