3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR30320-1G
APOSOR30320-1GEA 63 GBP
APOSOR30320-1G
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid
Formula: C₁₃H₁₄N₂O₄
MW: 262.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173707
CAS Number: 175278-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR