(2,4-Dioxo-1,2,3,4-tetrahydro-pyrimidin-5-ylamino)-fumaric acid dimethyl ester

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR303078-1G OR303078-500MG
APOSOR303078-1GEA 131 GBP
APOSOR303078-1G APOSOR303078-500MG
(2,4-Dioxo-1,2,3,4-tetrahydro-pyrimidin-5-ylamino)-fumaric acid dimethyl ester
(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester
Formula: C₁₀H₁₁N₃O₆
Storage Temperature: Ambient
MDL Number: MFCD22580727
CAS Number: 60458-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR