4-Chloroimidazo[4,5-c]pyridine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR302509-1G
APOSOR302509-1GEA 39.9 GBP
APOSOR302509-1G
4-Chloroimidazo[4,5-c]pyridine 98%
4-Chloroimidazo[4,5-c]pyridine
Formula: C₆H₄ClN₃
Melting Pt: 264 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689582
CAS Number: 2770-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR