2,4-Dichloro-6-methylbenzenesulphonyl chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2,4-Dichloro-6-methylbenzene-1-sulphonyl chloride

OR30237-1G OR30237-500MG OR30237-5G
APOSOR30237-1GEA 81.9 GBP
APOSOR30237-1G APOSOR30237-500MG APOSOR30237-5G
2,4-Dichloro-6-methylbenzenesulphonyl chloride
2,4-Dichloro-6-methylbenzenesulphonyl chloride
Formula: C₇H₅Cl₃O₂S
MW: 259.54
Storage Temperature: Ambient
MDL Number: MFCD00204179
CAS Number: 175278-26-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR