Avanafil

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR302290-1G OR302290-5G
APOSOR302290-1GEA 46.2 GBP
APOSOR302290-1G APOSOR302290-5G
Avanafil
Avanafil
Formula: C₂₃H₂₆ClN₇O₃
Storage Temperature: Ambient
MDL Number: MFCD11973638
CAS Number: 330784-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR