Ethyl 4,6-dichloro-2-methylnicotinate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR302196-25G
APOSOR302196-25GEA 131 GBP
APOSOR302196-25G
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Ethyl 4,6-dichloro-2-methylnicotinate
Formula: C₉H₉Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD05666193
CAS Number: 686279-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR