Pyrazolo[1,5-a]pyridine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR301332-1G
APOSOR301332-1GEA 70.4 GBP
APOSOR301332-1G
Pyrazolo[1,5-a]pyridine 97%
Pyrazolo[1,5-a]pyridine
Formula: C₇H₆N₂
MW: 118.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752622
CAS Number: 274-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR