4-Chloro-6-nitroquinoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR301169-1G
APOSOR301169-1GEA 209 GBP
APOSOR301169-1G
4-Chloro-6-nitroquinoline
4-Chloro-6-nitroquinoline
Formula: C₉H₅ClN₂O₂
MW: 208.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00498990
CAS Number: 13675-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR