Methyl 3-amino-2-bromobenzoate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR301136-1G OR301136-5G OR301136-10G
APOSOR301136-1GEA 18.9 GBP
APOSOR301136-1G APOSOR301136-5G APOSOR301136-10G
Methyl 3-amino-2-bromobenzoate
Methyl 3-amino-2-bromobenzoate
Formula: C₈H₈BrNO₂
Density: 1.583 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03844437
CAS Number: 106896-48-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR