3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR300379-1G OR300379-250MG
APOSOR300379-1GEA 397 GBP
APOSOR300379-1G APOSOR300379-250MG
3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine
3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine
Formula: C₆H₃BrClN₃
Storage Temperature: Ambient
MDL Number: MFCD14706800
CAS Number: 1245649-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR