6-Bromo-8-phenylimidazo[1,2-a]pyrazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR300223-250MG
APOSOR300223-250MGEA 188 GBP
APOSOR300223-250MG
6-Bromo-8-phenylimidazo[1,2-a]pyrazine
6-Bromo-8-phenylimidazo[1,2-a]pyrazine
Formula: C₁₂H₈BrN₃
MW: 274.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12033252
CAS Number: 1159815-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR