Ethyl-1,5-Dimethylpyrazole-3-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR29763-25G
APOSOR29763-25GEA 57.8 GBP
APOSOR29763-25G
Ethyl-1,5-Dimethylpyrazole-3-carboxylate
Ethyl-1,5-Dimethylpyrazole-3-carboxylate
Formula: C₈H₁₂N₂O₂
Boiling Pt: 154/10 mm °C
Melting Pt: 42…46 °C
MDL Number: MFCD00085071
CAS Number: 5744-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR