1,1,3,3-Tetramethylguanidine 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N,N,N',N'-Tetramethylguanidine

OR28879-100ML OR28879-500ML OR28879-1L
APOSOR28879-100MLEA 26.3 GBP
APOSOR28879-100ML APOSOR28879-500ML APOSOR28879-1L
1,1,3,3-Tetramethylguanidine 99%
1,1,3,3-Tetramethylguanidine
Formula: C₅H₁₃N₃
Boiling Pt: 162…163 °C
Density: 0.916 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR