3,4-Diaminobenzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR28812-500G
APOSOR28812-500GEA 167 GBP
APOSOR28812-500G
3,4-Diaminobenzoic acid 98%
3,4-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
Melting Pt: 208…210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR