Ethyl-6-bromohexanoate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate

OR28740-100G OR28740-500G
APOSOR28740-100GEA 49.4 GBP
APOSOR28740-100G APOSOR28740-500G
Ethyl-6-bromohexanoate
Ethyl 6-bromohexanoate
Formula: BrH₂C(CH₂)₄COOCH₂CH₃
Boiling Pt: 128…130 °C/16mmHg
Density: 1.254 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR